Assay Details
Target Gene Details
Entrez Gene ID: | 71045 |
Gene Name: | RIKEN cDNA 4933406I18 gene |
Gene Aliases: |
4933403D24Rik |
Location: |
Chr.7:114315479-114415161 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 4933406I18Rik | NR_029437.1 | |||
| AK016693.1 |

Genomic Map