Assay Details
Target Gene Details
Entrez Gene ID: | 20392 |
Gene Name: | sarcoglycan, epsilon |
Gene Aliases: |
e-SG |
Location: |
Chr.6:4674350-4747204 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sgce | NM_001130188.1 | 7 | 976 | NP_001123660.1 |
| NM_001130189.1 | 6 | 868 | NP_001123661.1 | |
| NM_001130190.1 | 6 | 868 | NP_001123662.1 | |
| NM_001130191.1 | 6 | 868 | NP_001123663.1 | |
| NM_011360.3 | 6 | 868 | NP_035490.3 | |
| AB117975.1 | 6 | 701 | BAD21207.1 | |
| AB200281.1 | 6 | 683 | BAE16175.1 | |
| AB200282.1 | 6 | 683 | BAE16176.1 | |
| AF031919.1 | 6 | 724 | AAC14020.1 | |
| AF103877.1 | 7 | 782 | AAF21895.1 | |
| AK076102.1 | 6 | 690 | BAC36184.1 | |
| AK132210.1 | 6 | 745 | BAE21034.1 | |
| BC012665.1 | 6 | 774 | AAH12665.1 |
More Information
Set Membership: |
|

Genomic Map