Assay Details
Target Gene Details
Entrez Gene ID: | 20216 |
Gene Name: | acyl-CoA synthetase medium-chain family member 3 |
Gene Aliases: |
Sa, Sah |
Location: |
Chr.7:119760883-119784896 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 15 - Intron 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Acsm3 | NM_016870.3 | NP_058566.3 | ||
| NM_212441.2 | NP_997606.2 | |||
| NM_212442.2 | NP_997607.2 | |||
| AB022340.2 | BAA37141.1 | |||
| AF068246.1 | AAC79656.1 | |||
| AK077247.1 | ||||
| AK165516.1 | BAE38232.1 | |||
| AY064696.1 | AAL40880.1 | |||
| BC015248.1 | AAH15248.1 |
Target Gene Details
Entrez Gene ID: | 71151 |
Gene Name: | exoribonuclease 2 |
Gene Aliases: |
4933424N09Rik, Exod1, mKIAA1504 |
Location: |
Chr.7:119783826-119794058 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Eri2 | NM_027698.5 | 9 | 3442 | NP_081974.3 |
| AK129376.1 | 7 | 3177 | BAC98186.1 | |
| AK137621.1 | 9 | 3389 | ||
| AK141507.1 | 9 | 3363 | ||
| AK165163.1 | 9 | 3352 | BAE38055.1 | |
| BC090961.1 | 9 | 3344 | AAH90961.1 |
More Information
Set Membership: |
|

Genomic Map