Assay Details
Target Gene Details
Entrez Gene ID: | 54399 |
Gene Name: | blocked early in transport 1 homolog (S. cerevisiae)-like |
Gene Aliases: |
2610021K23Rik, Gs15 |
Location: |
Chr.7:140853384-140856383 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Bet1l | NM_018742.5 | NP_061212.3 | ||
| BY292666.1 |
More Information
Set Membership: |
|

Genomic Map