Assay Details
Target Gene Details
Entrez Gene ID: | 11875 |
Gene Name: | ADP-ribosyltransferase 5 |
Gene Aliases: |
ARTC5, Yac-2 |
Location: |
Chr.7:102096879-102100306 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Art5 | NM_001291354.1 | NP_001278283.1 | ||
| NM_007491.2 | 2 | 499 | NP_031517.2 | |
| NR_111935.1 | ||||
| AK132854.1 | 2 | 500 | BAE21394.1 | |
| BC116823.1 | 2 | 311 | ||
| BC116849.1 | 2 | 311 | ||
| BC144825.1 | 2 | 311 | ||
| BC144826.1 | ||||
| BC144827.1 | 1 | 498 |
More Information
Set Membership: |
|

Genomic Map