Assay Details
Target Gene Details
Entrez Gene ID: | 11875 |
Gene Name: | ADP-ribosyltransferase 5 |
Gene Aliases: |
ARTC5, Yac-2 |
Location: |
Chr.7:102096879-102100306 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Art5 | NM_001291354.1 | NP_001278283.1 | ||
| NM_007491.2 | NP_031517.2 | |||
| NR_111935.1 | ||||
| AJ297547.1 | CAC50566.1 | |||
| AJ297548.1 | CAC50567.1 | |||
| AK132854.1 | BAE21394.1 | |||
| BC116823.1 | ||||
| BC116849.1 | ||||
| BC144825.1 | ||||
| BC144826.1 | ||||
| BC144827.1 |
More Information
Set Membership: |
|

Genomic Map