Assay Details
Target Gene Details
Entrez Gene ID: | 69785 |
Gene Name: | carcinoembryonic antigen-related cell adhesion molecule 13 |
Gene Aliases: |
1600012K03Rik, 1600025J19Rik |
Location: |
Chr.7:18009889-18019220 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ceacam13 | NM_027210.3 | NP_081486.1 | ||
| NM_028171.2 | NP_082447.1 | |||
| AK005430.1 | BAB24024.1 | |||
| AK005537.1 | BAB24108.1 | |||
| AK146045.1 | BAE26857.1 | |||
| BC099430.1 | AAH99430.1 | |||
| BY705748.1 |
More Information
Set Membership: |
|

Genomic Map