Assay Details
Target Gene Details
Entrez Gene ID: | 12611 |
Gene Name: | CCAAT/enhancer binding protein (C/EBP), gamma |
Gene Aliases: |
C/EBP[g], C/EBPgamma, GPE1-BP, Gpe1bp, Ig/EBP |
Location: |
Chr.7:35046422-35056590 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cebpg | NM_009884.3 | NP_034014.1 | ||
| AB012273.1 | BAA25311.1 | |||
| AK159544.1 | BAE35170.1 | |||
| BC011319.1 | AAH11319.1 | |||
| CJ092456.1 | ||||
| S80556.1 | 1 | 76 |
More Information
Set Membership: |
|

Genomic Map