Assay Details
Target Gene Details
Entrez Gene ID: | 12729 |
Gene Name: | chloride channel, nucleotide-sensitive, 1A |
Gene Aliases: |
2610036D06Rik, 2610100O04Rik, Clci, Clcni, ICLN |
Location: |
Chr.7:97696657-97720793 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Clns1a | NM_023671.2 | NP_076160.1 | ||
| AK011691.1 | ||||
| AK011789.1 | ||||
| AK031877.1 | BAC27588.1 | |||
| AK078050.1 | ||||
| AK133981.1 | ||||
| AK151356.1 | ||||
| AK151944.1 | ||||
| AK169259.1 | BAE41022.1 | |||
| AK205730.1 | ||||
| BC005575.1 | AAH05575.3 | |||
| CX236388.1 | ||||
| U53455.1 | AAB03317.1 |
More Information
Set Membership: |
|

Genomic Map