Assay Details
Target Gene Details
Entrez Gene ID: | 75538 |
Gene Name: | family with sequence similarity 71, member E1 |
Gene Aliases: |
0610007G24Rik, 1700021N13Rik, 1700021P22Rik, GARI-L5 |
Location: |
Chr.7:44496584-44501681 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fam71e1 | NM_028169.1 | 1 | 63 | NP_082445.1 |
| AB473729.1 | ||||
| AK006222.1 | BAB24467.1 | |||
| AK006229.1 | ||||
| AK018715.1 | 1 | 57 | ||
| BC049752.1 | ||||
| BC130017.1 |

Genomic Map