Assay Details
Target Gene Details
Entrez Gene ID: | 233899 |
Gene Name: | predicted gene 166 |
Gene Aliases: |
- |
Location: |
Chr.7:127582383-127588609 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gm166 | NM_001033040.3 | 4 | 904 | NP_001028212.1 |
| AK140121.1 | 4 | 905 | BAE24243.1 | |
| BC066021.1 | 4 | 485 |

Genomic Map