Assay Details
Target Gene Details
Entrez Gene ID: | 233899 |
Gene Name: | predicted gene 166 |
Gene Aliases: |
- |
Location: |
Chr.7:127582383-127588609 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gm166 | NM_001033040.3 | NP_001028212.1 | ||
| AK140121.1 | BAE24243.1 | |||
| BC066021.1 |

Genomic Map