Assay Details
Target Gene Details
Entrez Gene ID: | 14918 |
Gene Name: | guanylate cyclase 2d |
Gene Aliases: |
- |
Location: |
Chr.7:98440416-98477572 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gucy2d | NM_001130693.3 | 5 | 1549 | NP_001124165.1 |
More Information
Set Membership: |
|

Genomic Map