Assay Details
Target Gene Details
Entrez Gene ID: | 434232 |
Gene Name: | IQ motif containing K |
Gene Aliases: |
A230094G09Rik |
Location: |
Chr.7:118855755-118972652 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Iqck | NM_001081446.1 | 1 | 45 | NP_001074915.1 |
| AK132322.1 | 1 | 69 | ||
| AK138400.1 | 1 | 46 | BAE23645.1 | |
| AK143223.1 | 1 | 61 |
Target Gene Details
Entrez Gene ID: | 66356 |
Gene Name: | lysine rich nucleolar protein 1 |
Gene Aliases: |
2310008H09Rik, Tsg118 |
Location: |
Chr.7:118842217-118855998 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Knop1 | NM_001168218.1 | 1 | 180 | NP_001161690.1 |
| NM_001168219.1 | 1 | 180 | NP_001161691.1 | |
| NM_023197.3 | 1 | 180 | NP_075686.2 | |
| AF034580.1 | 2 | 349 | AAD01984.1 | |
| AK006113.1 | 1 | 184 | ||
| BB611983.1 | 1 | 69 |

Genomic Map