Assay Details
Target Gene Details
Entrez Gene ID: | 67425 |
Gene Name: | EPS8-like 1 |
Gene Aliases: |
2310051G19Rik, 4632407K17Rik, AW060268, DRC3, EPS8R1 |
Location: |
Chr.7:4460631-4480487 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Eps8l1 | NM_026146.4 | NP_080422.1 | ||
| AK009930.1 | ||||
| AK019490.2 | BAB31756.2 |
More Information
Set Membership: |
|

Genomic Map