Assay Details
Target Gene Details
Entrez Gene ID: | 70333 |
Gene Name: | CD3E antigen, epsilon polypeptide associated protein |
Gene Aliases: |
1190028F09, 2610103M17Rik, ASE-1, Ase1, CAST, PAF49 |
Location: |
Chr.7:19356007-19359483 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cd3eap | NM_145822.2 | 3 | 1865 | NP_665821.1 |
| AK075701.1 | 1 | 528 |
Target Gene Details
Entrez Gene ID: | 13870 |
Gene Name: | excision repair cross-complementing rodent repair deficiency, complementation group 1 |
Gene Aliases: |
Ercc-1 |
Location: |
Chr.7:19344422-19356524 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ercc1 | NM_007948.2 | NP_031974.2 | ||
| AK088271.1 | BAC40249.1 | |||
| AK136733.1 | BAE23112.1 | |||
| BC011224.1 | AAH11224.1 | |||
| BE949768.1 | ||||
| X07414.1 | CAA30310.1 |
More Information
Set Membership: |
|

Genomic Map