Assay Details
Target Gene Details
Entrez Gene ID: | 14113 |
Gene Name: | fibrillarin |
Gene Aliases: |
AL022665, FIB, FLRN, RNU3IP1 |
Location: |
Chr.7:28169748-28179269 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fbl | NM_007991.3 | NP_032017.2 | ||
| AK012620.1 | ||||
| AK146328.1 | BAE27083.1 | |||
| AV486126.1 | ||||
| BC003813.1 | AAH03813.1 | |||
| BC092274.1 | AAH92274.1 | |||
| BU511282.1 | ||||
| Z22593.1 | CAA80307.1 |
More Information
Set Membership: |
|

Genomic Map