Assay Details
Target Gene Details
Entrez Gene ID: | 15461 |
Gene Name: | Harvey rat sarcoma virus oncogene |
Gene Aliases: |
H-ras, Ha-ras, Harvey-ras, Hras-1, Hras1, Kras2, c-H-ras, c-Ha-ras, c-rasHa, ras |
Location: |
Chr.7:141189934-141194004 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hras | NM_001130443.1 | 3 | 455 | NP_001123915.1 |
| NM_001130444.1 | 3 | 455 | NP_001123916.1 | |
| NM_008284.2 | 3 | 455 | NP_032310.2 | |
| AK183914.1 | 2 | 214 | ||
| AY373386.1 | 2 | 249 | AAQ81319.1 | |
| BQ963436.1 | 3 | 444 | ||
| BU924724.1 | 1 | 91 | ||
| CK788792.1 | 3 | 455 |
More Information
Set Membership: |
|

Genomic Map