Assay Details
Target Gene Details
Entrez Gene ID: | 66245 |
Gene Name: | HSPA (heat shock 70kDa) binding protein, cytoplasmic cochaperone 1 |
Gene Aliases: |
1500019G21Rik |
Location: |
Chr.7:4660515-4685121 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hspbp1 | NM_024172.3 | 7 | 1107 | NP_077134.1 |
| AF338351.1 | 6 | 1045 | AAK11657.1 | |
| AK013263.1 | 7 | 1098 | BAB28756.1 | |
| AK088469.1 | 7 | 1083 | BAC40373.1 | |
| AK160128.1 | 7 | 1103 | ||
| AK184323.1 | ||||
| AK209519.1 | ||||
| BC014758.1 | 7 | 1107 | AAH14758.1 |

Genomic Map