Assay Details
Target Gene Details
Entrez Gene ID: | 18301 |
Gene Name: | FXYD domain-containing ion transport regulator 5 |
Gene Aliases: |
EF-8, Oit2, RIC |
Location: |
Chr.7:31032723-31042331 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fxyd5 | NM_001287213.1 | 1 | 84 | NP_001274142.1 |
| NM_001287217.1 | 1 | 84 | NP_001274146.1 | |
| NM_008761.4 | 1 | 84 | NP_032787.1 | |
| AI121044.1 | 1 | 84 | ||
| CA528582.1 | 1 | 75 |
More Information
Set Membership: |
|

Genomic Map