Assay Details
Target Gene Details
Entrez Gene ID: | 114640 |
Gene Name: | parathyroid hormone 2 |
Gene Aliases: |
Tifp39, Tip39 |
Location: |
Chr.7:45176565-45181838 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pth2 | NM_053256.2 | NP_444486.1 | ||
| AF399674.1 | AAK84862.1 | |||
| AY048587.1 | AAL06596.1 |

Genomic Map