Assay Details
Target Gene Details
Entrez Gene ID: | 69920 |
Gene Name: | polymerase (RNA) II (DNA directed) polypeptide I |
Gene Aliases: |
2810002B19Rik |
Location: |
Chr.7:30232074-30233387 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Polr2i | NM_027259.1 | NP_081535.1 | ||
| AB221537.1 | ||||
| AK012635.1 | ||||
| BC062812.1 | AAH62812.1 |
More Information
Set Membership: |
|

Genomic Map