Assay Details
Target Gene Details
Entrez Gene ID: | 70316 |
Gene Name: | NADH dehydrogenase (ubiquinone) 1, alpha/beta subcomplex, 1 |
Gene Aliases: |
2210401F17Rik, 2310039H15Rik, 2610003B19Rik, 8kDa, 9130423F15Rik, ACP, CI-SDAP, SDAP |
Location: |
Chr.7:122088044-122101877 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ndufab1 | NM_028177.3 | NP_082453.2 | ||
| AK009698.1 | BAB26446.1 | |||
| AK010307.1 | BAB26840.1 | |||
| AK011302.1 | BAB27528.1 | |||
| AK018688.1 | BAB31346.1 | |||
| AK018717.1 | BAB31363.1 | |||
| AK089103.1 | BAC40751.1 | |||
| AK151719.1 | BAE30638.1 | |||
| AK152995.1 | BAE31638.1 | |||
| AK167447.1 | BAE39533.1 | |||
| BC060951.1 | AAH60951.1 | |||
| BC092379.1 | AAH92379.1 |
More Information
Set Membership: |
|

Genomic Map