Assay Details
Target Gene Details
Entrez Gene ID: | 22342 |
Gene Name: | lin-7 homolog B (C. elegans) |
Gene Aliases: |
LIN-7B, MALS-2, Veli2 |
Location: |
Chr.7:45367891-45370564 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Lin7b | NM_011698.1 | NP_035828.1 | ||
| AF087694.1 | AAC78482.1 | |||
| AF173082.1 | AAD48501.1 | |||
| AK019299.1 | ||||
| BC031780.1 | AAH31780.1 |
Target Gene Details
Entrez Gene ID: | 50994 |
Gene Name: | metastasis associated gene 2 |
Gene Aliases: |
MAG |
Location: |
Chr.7:45366163-45367948 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mtag2 | NR_015480.1 | 2 | 601 | |
| NR_027802.1 | 1 | 630 | ||
| AK042966.1 | 2 | 601 | ||
| U88401.1 | 1 | 630 | AAB48499.1 |
More Information
Set Membership: |
|

Genomic Map