Assay Details
Target Gene Details
Entrez Gene ID: | 338352 |
Gene Name: | NEL-like 1 |
Gene Aliases: |
B230343H07Rik, l7R6 |
Location: |
Chr.7:49975350-50863289 on Build GRCm38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nell1 | NM_001037906.2 | 14 | 1529 | NP_001032995.1 |
| AY622226.1 | 14 | 1529 | AAV41488.1 |
Target Gene Details
Entrez Gene ID: | 73515 |
Gene Name: | NEL-like 1, opposite strand |
Gene Aliases: |
1700081H22Rik |
Location: |
Chr.7:50517309-50575884 on Build GRCm38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nell1os | NR_045928.1 | |||
| AK006968.1 |
More Information
Set Membership: |
|

Genomic Map