Assay Details
Target Gene Details
Entrez Gene ID: | 67994 |
Gene Name: | mitochondrial ribosomal protein S11 |
Gene Aliases: |
0710005I03Rik, C79873 |
Location: |
Chr.7:78783131-78792989 on Build GRCm38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mrps11 | NM_026498.2 | NP_080774.2 | ||
| AB049945.1 | BAB40998.1 | |||
| AK002993.1 | BAB22503.1 | |||
| AK152027.1 | BAE30886.1 | |||
| AK160207.1 | ||||
| AK166241.1 | BAE38655.1 | |||
| BC005701.1 | AAH05701.1 | |||
| BC028279.1 | AAH28279.1 |
More Information
Set Membership: |
|

Genomic Map