Assay Details
Target Gene Details
Entrez Gene ID: | 56189 |
Gene Name: | proline dehydrogenase (oxidase) 2 |
Gene Aliases: |
2510028N04Rik, 2510038B11Rik, MmPOX, MmPOX1, POX1 |
Location: |
Chr.7:30493586-30513402 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 6 - Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Prodh2 | NM_019546.5 | NP_062419.2 | ||
| AF222851.1 | AAQ13907.1 | |||
| AK011020.1 | ||||
| AK011039.1 | ||||
| AK050141.1 | BAC34090.1 | |||
| BC018182.1 | AAH18182.1 | |||
| U80019.1 | AAF21466.1 |
More Information
Set Membership: |
|

Genomic Map