Assay Details
Target Gene Details
Entrez Gene ID: | 68961 |
Gene Name: | phosphorylase kinase, gamma 2 (testis) |
Gene Aliases: |
1500017I02Rik |
Location: |
Chr.7:127573344-127583307 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Phkg2 | NM_026888.3 | 1 | 54 | NP_081164.2 |
| AK005277.1 | BAB23926.1 | |||
| AK079680.1 | 1 | 64 | ||
| AK089062.1 | ||||
| BC138913.1 | ||||
| BC145734.1 |
More Information
Set Membership: |
|

Genomic Map