Assay Details
Target Gene Details
Entrez Gene ID: | 233899 |
Gene Name: | predicted gene 166 |
Gene Aliases: |
- |
Location: |
Chr.7:127582383-127588609 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gm166 | NM_001033040.3 | 9 | 2336 | NP_001028212.1 |
| BC066021.1 | 9 | 1917 |
Target Gene Details
Entrez Gene ID: | 68961 |
Gene Name: | phosphorylase kinase, gamma 2 (testis) |
Gene Aliases: |
1500017I02Rik |
Location: |
Chr.7:127573344-127583307 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Phkg2 | NM_026888.3 | 9 | 993 | NP_081164.2 |
| AA198694.1 | 4 | 407 | ||
| AK005277.1 | 9 | 938 | BAB23926.1 | |
| BC138913.1 | 9 | 907 | ||
| BC145734.1 | 9 | 907 | ||
| BQ176174.1 |
More Information
Set Membership: |
|

Genomic Map