Assay Details
Target Gene Details
Entrez Gene ID: | 78405 |
Gene Name: | neurotrophin 5 |
Gene Aliases: |
2900040K06Rik, AI462899, NT-4, NT4, NT4/5, Ntf-5, Ntf4 |
Location: |
Chr.7:45413695-45417179 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ntf5 | NM_198190.1 | NP_937833.1 | ||
| BC052191.1 | AAH52191.1 |
More Information
Set Membership: |
|

Genomic Map