Assay Details
Target Gene Details
Entrez Gene ID: | 243880 |
Gene Name: | NLR family, pyrin domain containing 4A |
Gene Aliases: |
E330021B02, E330028A19Rik, NALP4, Nalp-eta, Nalp-ita, Nalp4a |
Location: |
Chr.7:26435113-26476142 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 5 - Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Nlrp4a | NM_172896.2 | NP_766484.1 | ||
AK054378.1 | ||||
AK087843.1 | BAC40024.1 | |||
AK135745.1 | ||||
AK136205.1 | ||||
AY355341.1 | AAR14738.1 | |||
AY596200.1 | AAU06321.1 | |||
BC141342.1 | ||||
BC141343.1 |
