Assay Details
Target Gene Details
Entrez Gene ID: | 243880 |
Gene Name: | NLR family, pyrin domain containing 4A |
Gene Aliases: |
E330021B02, E330028A19Rik, NALP4, Nalp-eta, Nalp-ita, Nalp4a |
Location: |
Chr.7:26435113-26476142 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Nlrp4a | NM_172896.2 | 6 | 2392 | NP_766484.1 |
AK054378.1 | 6 | 2283 | ||
AK087843.1 | 6 | 2393 | BAC40024.1 | |
AK135745.1 | 3 | 1206 | ||
AK136205.1 | 7 | 2368 | ||
AY355341.1 | 6 | 2393 | AAR14738.1 | |
AY596200.1 | 4 | 2089 | AAU06321.1 | |
BC141342.1 | 6 | 2292 | ||
BC141343.1 | 6 | 2292 |
