Assay Details
Target Gene Details
Entrez Gene ID: | 72434 |
Gene Name: | Ly6/Plaur domain containing 3 |
Gene Aliases: |
2310061G07Rik, C4.4a |
Location: |
Chr.7:24636570-24641118 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Lypd3 | NM_133743.1 | NP_598504.1 | ||
| AK009998.1 | ||||
| BC016549.1 | AAH16549.1 | |||
| BC094282.1 | AAH94282.1 |
More Information
Set Membership: |
|

Genomic Map