Assay Details
Target Gene Details
Entrez Gene ID: | 75099 |
Gene Name: | LysM, putative peptidoglycan-binding, domain containing 4 |
Gene Aliases: |
4930506D23Rik |
Location: |
Chr.7:67222544-67228468 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Lysmd4 | NM_175215.4 | NP_780424.1 | ||
| AK172203.1 | BAE42879.1 |

Genomic Map