Assay Details
Target Gene Details
Entrez Gene ID: | 210045 |
Gene Name: | NLR family, pyrin domain containing 4B |
Gene Aliases: |
AU022726, E330024M09, Nalp-gamma, Nalp4b |
Location: |
Chr.7:10687793-10730168 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nlrp4b | NM_172481.2 | NP_766069.1 | ||
| AK054426.1 | BAC35775.1 | |||
| AK162217.1 | BAE36795.1 | |||
| AY355342.1 | AAR14739.1 | |||
| AY596198.1 | AAU06319.1 | |||
| BC066795.1 | AAH66795.1 |

Genomic Map