Assay Details
Target Gene Details
Entrez Gene ID: | 83564 |
Gene Name: | NLR family, pyrin domain containing 4C |
Gene Aliases: |
Nalp-alpha, Nalp4c, Rnh2 |
Location: |
Chr.7:6045161-6105509 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nlrp4c | NM_031389.2 | NP_113566.2 | ||
| AF285581.1 | AAK31960.1 | |||
| AK133284.1 | ||||
| AK166817.1 | BAE39043.1 | |||
| AK166869.1 | BAE39081.1 | |||
| AY596194.1 | AAU06315.1 | |||
| BC120833.1 | AAI20834.1 |

Genomic Map