Assay Details
Target Gene Details
Entrez Gene ID: | 20085 |
Gene Name: | ribosomal protein S19 |
Gene Aliases: |
Dsk3 |
Location: |
Chr.7:24884376-24889806 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rps19 | NM_023133.1 | NP_075622.1 | ||
| AK012052.1 | ||||
| AK013524.1 | BAB28898.1 | |||
| AK018725.1 | BAB31370.1 | |||
| AK028244.1 | BAC25836.1 | |||
| BC034506.1 | AAH34506.1 | |||
| BC086775.1 | AAH86775.1 | |||
| BC086938.1 | AAH86938.1 |
More Information
Set Membership: |
|

Genomic Map