Assay Details
Target Gene Details
Entrez Gene ID: | 69748 |
Gene Name: | aldehyde dehydrogenase 16 family, member A1 |
Gene Aliases: |
2410004H02Rik, AI452234, mFLJ00209 |
Location: |
Chr.7:45141833-45154538 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Aldh16a1 | NM_145954.1 | NP_666066.1 | ||
| AK030673.1 | BAC27074.1 | |||
| AK144995.1 | BAE26175.1 | |||
| AK151691.1 | BAE30615.1 | |||
| AK161198.1 | BAE36234.1 | |||
| AK164374.1 | 1 | 38 | ||
| AK170276.1 | BAE41680.1 | |||
| AK220200.1 | BAD90125.1 | |||
| BC013548.1 | AAH13548.2 |
Target Gene Details
Entrez Gene ID: | 68845 |
Gene Name: | PIH1 domain containing 1 |
Gene Aliases: |
1110061L23Rik, 4933413A04Rik, Nop17 |
Location: |
Chr.7:45153835-45160065 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pih1d1 | NM_001278207.1 | NP_001265136.1 | ||
| NM_029406.4 | NP_083682.1 | |||
| AK004341.1 | BAB23269.1 | |||
| AK016797.1 | BAB30435.1 | |||
| AK089003.1 | BAC40694.1 | |||
| AK146039.1 | BAE26851.1 | |||
| AK152695.1 | BAE31425.1 |
More Information
Set Membership: |
|

Genomic Map