Assay Details
Target Gene Details
Entrez Gene ID: | 446099 |
Gene Name: | NLR family, pyrin domain containing 4E |
Gene Aliases: |
4930406H16Rik, Nalp-epsilon, Nalp4e, Nlrp4 |
Location: |
Chr.7:23301192-23400240 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nlrp4e |

Genomic Map