Assay Details
Target Gene Details
Entrez Gene ID: | 243937 |
Gene Name: | zinc finger protein 536 |
Gene Aliases: |
9630010P11Rik, Znf536, mKIAA0390 |
Location: |
Chr.7:37318023-37774177 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Zfp536 | NM_172385.2 | NP_759017.1 | ||
| AK030633.1 | BAC27055.1 | |||
| AK034508.1 | BAC28735.1 | |||
| AK122270.1 | BAC65552.1 | |||
| BC033594.1 | AAH33594.1 |
More Information
Set Membership: |
|

Genomic Map