Assay Details
Target Gene Details
Entrez Gene ID: | 24058 |
Gene Name: | single immunoglobulin and toll-interleukin 1 receptor (TIR) domain |
Gene Aliases: |
AI256711, TIR8 |
Location: |
Chr.7:141091175-141100610 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sigirr | NM_023059.3 | NP_075546.2 | ||
| AF113795.1 | AAF26200.1 | |||
| AF239957.1 | AAF44701.1 | |||
| AK133518.1 | BAE21700.1 | |||
| AK145889.1 | BAE26728.1 | |||
| BC010806.1 | AAH10806.1 | |||
| BC094069.1 | AAH94069.1 |
More Information
Set Membership: |
|

Genomic Map