Assay Details
Target Gene Details
Entrez Gene ID: | 320683 |
Gene Name: | zinc finger protein 629 |
Gene Aliases: |
9330199A09Rik, Znf629 |
Location: |
Chr.7:127607031-127615786 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Zfp629 | NM_177226.5 | NP_796200.1 | ||
| AK031161.1 | BAC27286.1 | |||
| AK034484.1 | BAC28725.1 | |||
| AK172933.1 | BAD32211.1 | |||
| BC035547.1 | AAH35547.1 | |||
| BC067053.1 | AAH67053.1 |
More Information
Set Membership: |
|

Genomic Map