Assay Details
Target Gene Details
Entrez Gene ID: | 233073 |
Gene Name: | U2 small nuclear RNA auxiliary factor 1-like 4 |
Gene Aliases: |
AA407033, AF419339, AI451269, AW553050, U2af26 |
Location: |
Chr.7:30563340-30565364 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 5 - Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| U2af1l4 | NM_170760.3 | NP_739566.1 | ||
| AF419339.1 | AAN63524.1 | |||
| AK180639.1 | ||||
| BC060972.1 | AAH60972.1 |
More Information
Set Membership: |
|

Genomic Map