Assay Details
Target Gene Details
Entrez Gene ID: | 12406 |
Gene Name: | serine (or cysteine) peptidase inhibitor, clade H, member 1 |
Gene Aliases: |
BERF-1, Cbp1, Cbp2, Hsp47, J6, Serpinh2, gp46 |
Location: |
Chr.7:99345375-99353239 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Serpinh1 | NM_001111043.1 | 5 | 1033 | NP_001104513.1 |
| NM_001111044.1 | 4 | 989 | NP_001104514.1 | |
| NM_001285776.1 | 5 | 1073 | NP_001272705.1 | |
| NM_009825.2 | 4 | 1160 | NP_033955.2 | |
| AK029080.1 | 5 | 951 | BAC26283.1 | |
| AK031457.1 | 4 | 1038 | BAC27413.1 | |
| AK076474.1 | 4 | 865 | BAC36360.1 | |
| AK077660.1 | 4 | 866 | BAC36935.1 | |
| AK077937.1 | 4 | 892 | BAC37075.1 | |
| AK077992.1 | ||||
| AK079460.1 | 4 | 865 | BAC37652.1 | |
| AK086519.1 | 4 | 865 | BAC39683.1 | |
| AK086523.1 | 4 | 865 | BAC39684.1 | |
| AK089993.1 | 4 | 867 | BAC41032.1 | |
| AK090103.1 | 4 | 867 | BAC41093.1 | |
| AK159697.1 | 4 | 867 | BAE35297.1 | |
| AK159998.1 | 4 | 867 | BAE35546.1 | |
| AK166000.1 | 4 | 865 | BAE38510.1 | |
| AK167403.1 | 4 | 865 | BAE39492.1 | |
| AK168442.1 | 4 | 868 | BAE40347.1 | |
| AK168870.1 | 4 | 843 | BAE40689.1 | |
| AK169422.1 | ||||
| BC085143.1 | 5 | 906 | AAH85143.1 | |
| J05609.1 | 5 | 900 | AAA03200.1 | |
| X60676.1 | 4 | 1015 | CAA43091.1 |
More Information
Set Membership: |
|

Genomic Map