Assay Details
Target Gene Details
Entrez Gene ID: | 101883 |
Gene Name: | IGF-like family receptor 1 |
Gene Aliases: |
F630003I17Rik, Tmem149 |
Location: |
Chr.7:30565427-30567962 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Igflr1 | NM_145580.2 | 3 | 359 | NP_663555.2 |
| AK154129.1 | 3 | 360 | BAE32394.1 | |
| BC019733.1 | 2 | 403 | AAH19733.1 |
More Information
Set Membership: |
|

Genomic Map