Assay Details
Target Gene Details
Entrez Gene ID: | 54006 |
Gene Name: | deformed epidermal autoregulatory factor 1 (Drosophila) |
Gene Aliases: |
AU042387, C230009B13Rik, NUDR |
Location: |
Chr.7:141297176-141338870 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Deaf1 | NM_001282073.1 | NP_001269002.1 | ||
| BC046399.1 | AAH46399.1 |
Target Gene Details
Entrez Gene ID: | 71448 |
Gene Name: | transmembrane protein 80 |
Gene Aliases: |
5530601I19Rik |
Location: |
Chr.7:141327743-141337156 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tmem80 | NM_001141950.1 | NP_001135422.1 | ||
| NM_027797.3 | 3 | 335 | NP_082073.1 | |
| NR_024509.1 | 3 | 335 | ||
| AK017460.1 | 3 | 336 | BAB30753.1 | |
| AK134821.1 | BAE22300.1 | |||
| AK148367.1 | 3 | 306 | ||
| AK169083.1 | BAE40867.1 | |||
| BC022756.1 | AAH22756.1 |
More Information
Set Membership: |
|

Genomic Map