Assay Details
Target Gene Details
Entrez Gene ID: | 22228 |
Gene Name: | uncoupling protein 2 (mitochondrial, proton carrier) |
Gene Aliases: |
Slc25a8 |
Location: |
Chr.7:100493358-100499630 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ucp2 | NM_011671.5 | 2 | 191 | NP_035801.3 |
| AK134116.1 | 2 | 192 | BAE22021.1 | |
| AK142687.1 | 2 | 192 | BAE25160.1 | |
| AK165245.1 | 2 | 192 | ||
| AK172307.1 | 2 | 186 | ||
| BC012697.1 | 2 | 190 | AAH12697.1 | |
| BC012967.1 | 1 | 92 | AAH12967.1 | |
| BY299894.1 | 2 | 214 | ||
| U69135.1 | 2 | 119 | AAB17666.1 | |
| U94593.1 | 2 | 112 | AAB53092.1 |
More Information
Set Membership: |
|

Genomic Map