Assay Details
Target Gene Details
Entrez Gene ID: | 22228 |
Gene Name: | uncoupling protein 2 (mitochondrial, proton carrier) |
Gene Aliases: |
Slc25a8 |
Location: |
Chr.7:100493358-100499630 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ucp2 | NM_011671.5 | 7 | 1052 | NP_035801.3 |
| AF111998.1 | 5 | 755 | AAD17198.1 | |
| AF111999.1 | 5 | 749 | AAD17199.1 | |
| AK134116.1 | 7 | 1053 | BAE22021.1 | |
| AK142687.1 | 7 | 1053 | BAE25160.1 | |
| AK165245.1 | 7 | 1053 | ||
| BC012697.1 | 7 | 1051 | AAH12697.1 | |
| BC012967.1 | 6 | 953 | AAH12967.1 | |
| U69135.1 | 7 | 980 | AAB17666.1 | |
| U94593.1 | 7 | 973 | AAB53092.1 |
More Information
Set Membership: |
|

Genomic Map