Assay Details
Target Gene Details
Entrez Gene ID: | 22229 |
Gene Name: | uncoupling protein 3 (mitochondrial, proton carrier) |
Gene Aliases: |
AI645527, Slc25a9, UCP-3 |
Location: |
Chr.7:100472991-100486432 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ucp3 | NM_009464.3 | NP_033490.1 | ||
| AB008216.1 | BAA33502.1 | |||
| AB010742.1 | BAA25697.1 | |||
| AF019883.1 | AAB71543.1 | |||
| AF030164.1 | AAD01892.1 | |||
| AF032902.1 | AAB87084.1 | |||
| AF053352.1 | AAC28328.1 | |||
| BC139430.1 | ||||
| BC139431.1 |
More Information
Set Membership: |
|

Genomic Map