Assay Details
Target Gene Details
Entrez Gene ID: | 68267 |
Gene Name: | solute carrier family 25 (mitochondrial carrier, glutamate), member 22 |
Gene Aliases: |
1300006L01Rik, AI060884, Gc1 |
Location: |
Chr.7:141429749-141437887 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc25a22 | NM_001177576.1 | NP_001171047.1 | ||
| NM_026646.3 | NP_080922.1 | |||
| AK010193.1 | BAB26760.1 | |||
| AK018760.1 | ||||
| AK046874.1 | BAC32903.1 | |||
| AK048389.1 | BAC33318.1 | |||
| AK049313.1 | BAC33676.1 | |||
| AK155276.1 | BAE33159.1 | |||
| AK158985.1 | 1 | 1780 | ||
| AK162400.1 | BAE36895.1 | |||
| BC037949.1 | AAH37949.1 | |||
| BC050887.1 | AAH50887.1 | |||
| BY296753.1 |
More Information
Set Membership: |
|

Genomic Map